man

Test

CAS RN 1373920-78-5

CH3OOClFFFF

CAS RN 1373920-78-5

IUPAC Name: 2-fluoro-5-methoxy-3-(trifluoromethyl)benzoyl chloride
Synonyms: 2-fluoro-5-methoxy-3-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 256.58
Molecular Formula: C9H5ClF4O2
InChi Key: FKJAOSVOAJPKGX-UHFFFAOYSA-N
SMILES: COC1=CC(=C(F)C(=C1)C(Cl)=O)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results