man

Test

CAS RN 1373920-92-3

CH3NFFFF

CAS RN 1373920-92-3

IUPAC Name: 4-fluoro-3-methyl-5-(trifluoromethyl)benzonitrile
Synonyms: 4-fluoro-3-methyl-5-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 203.14
Molecular Formula: C9H5F4N
InChi Key: LHJDUSUXBSSXAY-UHFFFAOYSA-N
SMILES: CC1=C(F)C(=CC(=C1)C#N)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results