man

Test

CAS RN 137553-42-5

IFN

CAS RN 137553-42-5

IUPAC Name: 2-fluoro-4-iodobenzonitrile
Synonyms: 2-fluoro-4-iodobenzonitrile
Molecular Weight (g/mol): 247.01
Molecular Formula: C7H3FIN
InChi Key: WHQVXHBSTRFRCE-UHFFFAOYSA-N
SMILES: FC1=C(C=CC(I)=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results