man

Test

CAS RN 1380918-98-8

CFFZnBr

CAS RN 1380918-98-8

IUPAC Name: 2,6-difluorobenzen-1-ide; bromozincylium
Synonyms: 2,6-difluorobenzen-1-ide; bromozincylium
Molecular Weight (g/mol): 258.37
Molecular Formula: C6H3BrF2Zn
InChi Key: PYSMUDJGPPCOFC-UHFFFAOYSA-M
SMILES: [Zn+]Br.FC1=CC=CC(F)=[C-]1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results