CAS RN 13826-95-4
CAS RN 13826-95-4
IUPAC Name:
dilithium(1+) tetrabromonickeldiuide
Synonyms:
dilithium(1+) tetrabromonickelate(2-)
Molecular Weight (g/mol):
392.19
Molecular Formula:
Br4Li2Ni
InChi Key:
AVJDNPHXGKKFIA-UHFFFAOYSA-J
SMILES:
[Li+].[Li+].Br[Ni--](Br)(Br)Br
1
–
1
of
1
results
1
Thermo Scientific Acros Dilithium tetrabromonickelate(II), 0.5M solution in THF, AcroSeal™
CAS: 13826-95-4 Molecular Formula: Br4Li2Ni Molecular Weight (g/mol): 392.19 InChI Key: AVJDNPHXGKKFIA-UHFFFAOYSA-J Synonym: di-lithium tetrabromonickelate ii,agn-pc-0nf0dx,lithium tetrabromonickelate ii solution,dilithium tetrabromonickelate ii solution PubChem CID: 11047451 IUPAC Name: dilithium;tetrabromonickel(2-) SMILES: [Li+].[Li+].[Ni-2](Br)(Br)(Br)Br