CAS RN 13842-67-6
CAS RN 13842-67-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Terbium(III) sulfate octahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
CAS: 13842-67-6 MDL Number: MFCD00149872
2
Terbium(III) sulfate octahydrate, 99.995% (metals basis), Thermo Scientific™
CAS: 13842-67-6 Molecular Formula: Tb2(SO4)3·8H2O MDL Number: MFCD00149872