man

Test

CAS RN 1392494-64-2

CAS RN 1392494-64-2

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results