man

Test

CAS RN 140158-50-5

CH3H3CCH3H2NOHOOHNO

CAS RN 140158-50-5

IUPAC Name: 2-amino-3-(5-tert-butyl-3-oxo-2,3-dihydro-1,2-oxazol-4-yl)propanoic acid
Synonyms: 2-amino-3-(5-tert-butyl-3-oxo-2H-1,2-oxazol-4-yl)propanoic acid
Molecular Weight (g/mol): 228.25
Molecular Formula: C10H16N2O4
InChi Key: PIXJURSCCVBKRF-UHFFFAOYNA-N
SMILES: CC(C)(C)C1=C(CC(N)C(O)=O)C(=O)NO1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results