man

Test

CAS RN 14024-48-7

OOOOH3CH3CCH3CH3Co

CAS RN 14024-48-7

IUPAC Name: 2,4,8,10-tetramethyl-1λ³,5λ³,7λ³,11λ³-tetraoxa-6-cobaltaspiro[5.5]undeca-1,4,7,10-tetraene-6,6-bis(ylium)-3,9-diide
Synonyms: 2,4,8,10-tetramethyl-1λ³,5λ³,7λ³,11λ³-tetraoxa-6-cobaltaspiro[5.5]undeca-1,4,7,10-tetraene-6,6-bis(ylium)-3,9-diide
Molecular Weight (g/mol): 257.15
Molecular Formula: C10H14CoO4
InChi Key: BKFAZDGHFACXKY-UHFFFAOYSA-N
SMILES: CC1=[O][Co++]2([O]=C(C)[CH-]1)[O]=C(C)[CH-]C(C)=[O]2
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)