accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 14024-63-6

OOOOH3CH3CCH3CH3Zn

CAS RN 14024-63-6

IUPAC Name: zinc(2+) bis(2,4-dioxopentan-3-ide)
Synonyms: zinc(2+) diacetylacetonate
Molecular Weight (g/mol): 263.6
Molecular Formula: C10H14O4Zn
InChi Key: NHXVNEDMKGDNPR-UHFFFAOYSA-N
SMILES: [Zn++].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products