accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 141-03-7

H3COOOOCH3

CAS RN 141-03-7

IUPAC Name: 1,4-dibutyl butanedioate
Synonyms: butanedioic acid, dibutyl ester
Molecular Weight (g/mol): 230.3
Molecular Formula: C12H22O4
InChi Key: YUXIBTJKHLUKBD-UHFFFAOYSA-N
SMILES: CCCCOC(=O)CCC(=O)OCCCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products