CAS RN 141801-26-5
CAS RN 141801-26-5
IUPAC Name:
2-({1-[2-amino-3-(4-hydroxyphenyl)propanoyl]pyrrolidin-2-yl}formamido)-N-(1-carbamoyl-2-phenylethyl)-3-phenylpropanamide
Synonyms:
endomorphin-2
Molecular Weight (g/mol):
571.68
Molecular Formula:
C32H37N5O5
InChi Key:
XIJHWXXXIMEHKW-UHFFFAOYNA-N
SMILES:
NC(CC1=CC=C(O)C=C1)C(=O)N1CCCC1C(=O)NC(CC1=CC=CC=C1)C(=O)NC(CC1=CC=CC=C1)C(N)=O