man

Test

CAS RN 14235-81-5

NH2CH

CAS RN 14235-81-5

IUPAC Name: 4-ethynylaniline
Synonyms: 4-ethynylaniline
Molecular Weight (g/mol): 117.15
Molecular Formula: C8H7N
InChi Key: JXYITCJMBRETQX-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C#C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)