man

Test

CAS RN 1427451-43-1

H3COOBrF

CAS RN 1427451-43-1

IUPAC Name: methyl 2-(3-bromo-2-fluorophenyl)acetate
Synonyms: methyl 2-(3-bromo-2-fluorophenyl)acetate
Molecular Weight (g/mol): 247.06
Molecular Formula: C9H8BrFO2
InChi Key: ALPXGXOFMZOFAG-UHFFFAOYSA-N
SMILES: COC(=O)CC1=C(F)C(Br)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results