CAS RN 1428-67-7
CAS RN 1428-67-7
IUPAC Name:
2,3-bis(4-hydroxyphenyl)propanenitrile
Synonyms:
diarylpropionitrile
Molecular Weight (g/mol):
239.27
Molecular Formula:
C15H13NO2
InChi Key:
GHZHWDWADLAOIQ-UHFFFAOYNA-N
SMILES:
OC1=CC=C(CC(C#N)C2=CC=C(O)C=C2)C=C1
1
–
1
of
1
results
1
DPN, Tocris Bioscience™
CAS: 1428-67-7 Molecular Formula: C15H13NO2 Molecular Weight (g/mol): 239.274 InChI Key: GHZHWDWADLAOIQ-UHFFFAOYSA-N Synonym: diarylpropionitrile,2,3-bis 4-hydroxyphenyl propionitrile,2,3-bis 4-hydroxyphenyl propanenitrile,2,3-bis 4-hydroxyphenyl-propionitrile,2,3-bis p-hydroxyphenyl propionitrile,dpn compound,benzenepropanenitrile,4-hydroxy-a-4-hydroxyphenyl,diarylpropionitril,2,3-bis p-hydroxyphenyl-propionitrile,propionitrile, 2,3-bis p-hydroxyphenyl PubChem CID: 102614 ChEBI: CHEBI:63949 IUPAC Name: 2,3-bis(4-hydroxyphenyl)propanenitrile SMILES: C1=CC(=CC=C1CC(C#N)C2=CC=C(C=C2)O)O