man

Test

CAS RN 14328-51-9

OOHH2N

CAS RN 14328-51-9

IUPAC Name: 2-amino-2-cyclohexylacetic acid
Synonyms: cyclohexylglycine
Molecular Weight (g/mol): 157.21
Molecular Formula: C8H15NO2
InChi Key: WAMWSIDTKSNDCU-UHFFFAOYNA-N
SMILES: NC(C1CCCCC1)C(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results