CAS RN 143360-00-3
CAS RN 143360-00-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris™ SMIP 004
SKP2 (S-phase kinase-associated protein 2) inhibitor; upregulates p27 and arrests cell cycle in G1 phase