CAS RN 143664-11-3
CAS RN 143664-11-3
IUPAC Name:
N-{4-[2-(6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-yl)ethyl]phenyl}-5-methoxy-9-oxo-9,10-dihydroacridine-4-carboxamide
Synonyms:
N-{4-[2-(6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)ethyl]phenyl}-5-methoxy-9-oxo-10H-acridine-4-carboxamide
Molecular Weight (g/mol):
563.65
Molecular Formula:
C34H33N3O5
InChi Key:
OSFCMRGOZNQUSW-UHFFFAOYSA-N
SMILES:
COC1=C2NC3=C(C=CC=C3C(=O)C2=CC=C1)C(=O)NC1=CC=C(CCN2CCC3=CC(OC)=C(OC)C=C3C2)C=C1
1
–
1
of
1
results
1
Elacridar, Thermo Scientific Chemicals
CAS: 143664-11-3 Molecular Formula: C34H33N3O5 Molecular Weight (g/mol): 563.65 InChI Key: OSFCMRGOZNQUSW-UHFFFAOYSA-N IUPAC Name: N-{4-[2-(6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-yl)ethyl]phenyl}-5-methoxy-9-oxo-9,10-dihydroacridine-4-carboxamide SMILES: COC1=C2NC3=C(C=CC=C3C(=O)C2=CC=C1)C(=O)NC1=CC=C(CCN2CCC3=CC(OC)=C(OC)C=C3C2)C=C1