man

Test

CAS RN 144284-25-3

HOFFF

CAS RN 144284-25-3

IUPAC Name: (2,4,5-trifluorophenyl)methanol
Synonyms: (2,4,5-trifluorophenyl)methanol
Molecular Weight (g/mol): 162.11
Molecular Formula: C7H5F3O
InChi Key: NRXZCCOHXZFHBV-UHFFFAOYSA-N
SMILES: OCC1=CC(F)=C(F)C=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results