accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1445-39-2

NH2NIN

CAS RN 1445-39-2

IUPAC Name: 5-iodopyrimidin-2-amine
Synonyms: 5-iodopyrimidin-2-amine
Molecular Weight (g/mol): 221
Molecular Formula: C4H4IN3
InChi Key: HAFKCGZQRIIADX-UHFFFAOYSA-N
SMILES: NC1=NC=C(I)C=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products