CAS RN 14483-17-1
CAS RN 14483-17-1
IUPAC Name:
praseodymium(3+) pentahydrate trinitrate
Synonyms:
praseodymium(3+) pentahydrate trinitrate
Molecular Weight (g/mol):
417
Molecular Formula:
H10N3O14Pr
InChi Key:
PZVBXGDSTWEFMC-UHFFFAOYSA-N
SMILES:
O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O