man

Test

CAS RN 1448858-83-0

ONHONOOPOOHOPOHOOHOHOHINaNaNa

CAS RN 1448858-83-0

IUPAC Name: [({[3,4-dihydroxy-5-(5-iodo-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)oxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy]phosphonic acid trisodium
Synonyms: {[3,4-dihydroxy-5-(5-iodo-2,4-dioxo-3H-pyrimidin-1-yl)oxolan-2-yl]methoxy(hydroxy)phosphoryl}oxyphosphonic acid trisodium
Molecular Weight (g/mol): 599.03
Molecular Formula: C9H13IN2Na3O12P2
InChi Key: WAJPEYKEXKCDGT-UHFFFAOYNA-N
SMILES: [Na].[Na].[Na].OC1C(O)C(OC1COP(O)(=O)OP(O)(O)=O)N1C=C(I)C(=O)NC1=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results