accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 144978-12-1

CH3OOONOOCH3H3CCH3

CAS RN 144978-12-1

IUPAC Name: 1-tert-butyl 2-ethyl 5-oxopyrrolidine-1,2-dicarboxylate
Synonyms: 1-tert-butyl 2-ethyl 5-oxopyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 257.29
Molecular Formula: C12H19NO5
InChi Key: YWWWGFSJHCFVOW-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCC(=O)N1C(=O)OC(C)(C)C

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products