accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 14533-63-2

NaCH3SOOO

CAS RN 14533-63-2

IUPAC Name: sodium propane-1-sulfonate
Synonyms: sodium propane-1-sulfonate
Molecular Weight (g/mol): 146.14
Molecular Formula: C3H7NaO3S
InChi Key: NPAWNPCNZAPTKA-UHFFFAOYSA-M
SMILES: [Na+].CCCS([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products