man

Test

CAS RN 1464-42-2

SeNH2OHOH3C

CAS RN 1464-42-2

IUPAC Name: (2S)-2-amino-4-(methylselanyl)butanoic acid
Synonyms: L-selenomethionine
Molecular Weight (g/mol): 196.12
Molecular Formula: C5H11NO2Se
InChi Key: RJFAYQIBOAGBLC-BYPYZUCNSA-N
SMILES: C[Se]CC[C@H](N)C(O)=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)