CAS RN 146616-66-2
CAS RN 146616-66-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ BDY FL, SE
Green BDY (BODIPY™ or boron-dipyrromethene) fluorescent dye for the labeling of amines; supplied as NHS ester