CAS RN 14692-29-6
CAS RN 14692-29-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Strontium acetate hemihydrate, Reagent Grade, Thermo Scientific Chemicals
CAS: 14692-29-6 MDL Number: MFCD00198064
2
Strontium acetate hemihydrate, 98%, Thermo Scientific Chemicals
CAS: 14692-29-6 Molecular Formula: Sr(OOCCH3)2·0.5H2O MDL Number: MFCD00198064 Synonym: Acetic acid strontium salt