man

Test

CAS RN 147098-18-8

CH3H3CNNOOOHOHFNH3CSOOCH3Na

CAS RN 147098-18-8

IUPAC Name: sodium 7-[4-(4-fluorophenyl)-2-(N-methylmethanesulfonamido)-6-(propan-2-yl)pyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate
Synonyms: sodium 7-[4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethanesulfonamido)pyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate
Molecular Weight (g/mol): 503.52
Molecular Formula: C22H27FN3NaO6S
InChi Key: RGEBGDYYHAFODH-UHFFFAOYNA-M
SMILES: [Na+].CC(C)C1=NC(=NC(C2=CC=C(F)C=C2)=C1C=CC(O)CC(O)CC([O-])=O)N(C)S(C)(=O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results