CAS RN 147523-65-7
CAS RN 147523-65-7
IUPAC Name:
N-(4-bromophenyl)-3-oxo-4,5-diphenylpyrazolidine-1-carboxamide
Synonyms:
N-(4-bromophenyl)-3-oxo-4,5-diphenylpyrazolidine-1-carboxamide
Molecular Weight (g/mol):
436.31
Molecular Formula:
C22H18BrN3O2
InChi Key:
LMUQHXHWJWQXSD-UHFFFAOYNA-N
SMILES:
BrC1=CC=C(NC(=O)N2NC(=O)C(C2C2=CC=CC=C2)C2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
LY 288513, Tocris Bioscience™
CAS: 147523-65-7 Molecular Formula: C22H18BrN3O2 Molecular Weight (g/mol): 436.309 InChI Key: LMUQHXHWJWQXSD-PMACEKPBSA-N Synonym: unii-470gwp2ca0,4s,5r-n-4-bromophenyl-3-oxo-4,5-diphenyl-1-pyrazolidinecarboxamide,4s,5r-n-4-bromophenyl-3-oxo-4,5-diphenylpyrazolidine-1-carboxamide,maybridge4_000682,1-4,5-diphenyl-3-pyrazolidinone,1-pyrazolidinecarboxamide, n-4-bromophenyl-3-oxo-4,5-diphenyl-, 4s,5r,4s,5r-3-oxo-4,5-diphenyl-pyrazolidine-1-carboxylic acid 4-bromo-phenyl-amide,3-oxo-4,5-diphenyl-pyrazolidine-1-carboxylic acid 4-bromo-phenyl-amide ly288513 PubChem CID: 2802894 IUPAC Name: (4S,5R)-N-(4-bromophenyl)-3-oxo-4,5-diphenylpyrazolidine-1-carboxamide SMILES: C1=CC=C(C=C1)C2C(N(NC2=O)C(=O)NC3=CC=C(C=C3)Br)C4=CC=CC=C4