man

Test

CAS RN 147539-41-1

H3CNHNOOCH3H3CCH3

CAS RN 147539-41-1

IUPAC Name: tert-butyl 4-(methylamino)piperidine-1-carboxylate
Synonyms: tert-butyl 4-(methylamino)piperidine-1-carboxylate
Molecular Weight (g/mol): 214.31
Molecular Formula: C11H22N2O2
InChi Key: CZYUGTLMFHDODF-UHFFFAOYSA-N
SMILES: CNC1CCN(CC1)C(=O)OC(C)(C)C

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results