man

Test

CAS RN 148687-76-7

NHOFFFFFFHCl

CAS RN 148687-76-7

IUPAC Name: 3-{[3,5-bis(trifluoromethyl)phenyl]methoxy}-2-phenylpiperidine hydrochloride
Synonyms: 3-{[3,5-bis(trifluoromethyl)phenyl]methoxy}-2-phenylpiperidine hydrochloride
Molecular Weight (g/mol): 439.83
Molecular Formula: C20H20ClF6NO
InChi Key: DYEUTIUITGHIEO-UHFFFAOYNA-N
SMILES: Cl.FC(F)(F)C1=CC(=CC(COC2CCCNC2C2=CC=CC=C2)=C1)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results