man

Test

CAS RN 14914-84-2

H2OH2OH2OH2OH2OH2OHoClClCl

CAS RN 14914-84-2

IUPAC Name: trichloroholmium hexahydrate
Synonyms: trichloroholmium hexahydrate
Molecular Weight (g/mol): 379.37
Molecular Formula: Cl3H12HoO6
InChi Key: BUQFCXABQYNXLP-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.Cl[Ho](Cl)Cl
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results