CAS RN 14917-55-6
CAS RN 14917-55-6
IUPAC Name:
2-methoxy-6-methyloxane-3,4,5-triol
Synonyms:
(-)-fucose 6-deoxy-L-β-galactose rhodeose isodulcit mannomethylose, L- L-(-)-fucose l-(+)-rhamnose monohydrate uridine 5-pyrophosphate L-galactose, 6-deoxy- L-fuc
Molecular Weight (g/mol):
178.18
Molecular Formula:
C7H14O5
InChi Key:
OHWCAVRRXKJCRB-UHFFFAOYNA-N
SMILES:
COC1OC(C)C(O)C(O)C1O
1
–
1
of
1
results
1
Methyl alpha-L-rhamnopyranoside, 98%, Thermo Scientific Chemicals
CAS: 14917-55-6 Molecular Formula: C7H14O5 Molecular Weight (g/mol): 178.184 MDL Number: MFCD00067654 InChI Key: OHWCAVRRXKJCRB-PAMBMQIZSA-N Synonym: methyl alpha-l-rhamnopyranoside,methyl-alpha-l-rhamnopyranoside,1-o-methyl-alpha-rhamnose,methyl 6-deoxy-alpha-l-mannopyranoside,methyl a-l-rhamnopyranoside,rao,methyl rhamnopyranoside,2r,3r,4r,5r,6s-2-methoxy-6-methyloxane-3,4,5-triol,2r,3r,4r,5r,6s-2-methoxy-6-methyltetrahydro-2h-pyran-3,4,5-triol,methyl-a-l-rhamnopyranoside PubChem CID: 84695 IUPAC Name: (2R,3R,4R,5R,6S)-2-methoxy-6-methyloxane-3,4,5-triol SMILES: CC1C(C(C(C(O1)OC)O)O)O