CAS RN 149507-36-8
CAS RN 149507-36-8
IUPAC Name:
[4-methoxy-3-(trifluoromethyl)phenyl]boronic acid
Synonyms:
4-methoxy-3-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol):
219.95
Molecular Formula:
C8H8BF3O3
InChi Key:
BUSMBMGODABSIN-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1C(F)(F)F)B(O)O
1
–
1
of
1
results
1
4-Methoxy-3-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
CAS: 149507-36-8 Molecular Formula: C8H8BF3O3 Molecular Weight (g/mol): 219.954 MDL Number: MFCD07363790 InChI Key: BUSMBMGODABSIN-UHFFFAOYSA-N Synonym: 4-methoxy-3-trifluoromethyl phenyl boronic acid,4-methoxy-3-trifluoromethylphenylboronic acid,3-trifluoromethyl-4-methoxy-phenylboronic acid,4-methoxy-3-trifluoromethylphenyl boronic acid,4-methoxy-3-trifluoromethyl benzeneboronic acid,4-methoxy-3-trifluoromethyl phenylboronic acid,boronic acid, 4-methoxy-3-trifluoromethyl phenyl,acmc-1c5ik PubChem CID: 17750052 IUPAC Name: [4-methoxy-3-(trifluoromethyl)phenyl]boronic acid SMILES: B(C1=CC(=C(C=C1)OC)C(F)(F)F)(O)O