accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1496-40-8

NFFFH2N

CAS RN 1496-40-8

IUPAC Name: 2-(piperidin-1-yl)-5-(trifluoromethyl)aniline
Synonyms: 2-(piperidin-1-yl)-5-(trifluoromethyl)aniline
Molecular Weight (g/mol): 244.26
Molecular Formula: C12H15F3N2
InChi Key: BERRRZOJDANPHE-UHFFFAOYSA-N
SMILES: NC1=C(C=CC(=C1)C(F)(F)F)N1CCCCC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products