CAS RN 1498-42-6
CAS RN 1498-42-6
IUPAC Name:
ethyl dichlorophosphinite
Synonyms:
ethyl dichlorophosphinite
Molecular Weight (g/mol):
146.93
Molecular Formula:
C2H5Cl2OP
InChi Key:
PVCINRPAXRJLEP-UHFFFAOYSA-N
SMILES:
CCOP(Cl)Cl
1
–
1
of
1
results
1
Ethyl phosphorodichloridite, 98%, Thermo Scientific Chemicals
CAS: 1498-42-6 Molecular Formula: C2H5Cl2OP Molecular Weight (g/mol): 146.93 MDL Number: MFCD00000525 InChI Key: PVCINRPAXRJLEP-UHFFFAOYSA-N Synonym: ethyl dichlorophosphite,ethyl phosphorodichloridite,ethyl dichlorophosphinite,ethylphosphorodichloridite,ethyl phosphorodichloridoite,acmc-20akws,dichloroethoxyphosphine,dichloro ethoxy phosphane,dichloro ethoxy phosphine,c2h5opcl2 PubChem CID: 73905 IUPAC Name: dichloro(ethoxy)phosphane SMILES: CCOP(Cl)Cl