man

Test

CAS RN 1498-54-0

H3CNCH3PClClO

CAS RN 1498-54-0

IUPAC Name: (diethylamino)phosphonoyl dichloride
Synonyms: diethylaminophosphonoyl dichloride
Molecular Weight (g/mol): 190
Molecular Formula: C4H10Cl2NOP
InChi Key: FXKRFCBTXGJQPY-UHFFFAOYSA-N
SMILES: CCN(CC)P(Cl)(Cl)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results