CAS RN 149947-19-3
CAS RN 149947-19-3
IUPAC Name:
(2E)-3-(4-bromo-2-fluorophenyl)prop-2-enoate
Synonyms:
(2E)-3-(4-bromo-2-fluorophenyl)prop-2-enoate
Molecular Weight (g/mol):
244.04
Molecular Formula:
C9H5BrFO2
InChi Key:
SVJGKQYKYNDYMU-DUXPYHPUSA-M
SMILES:
[O-]C(=O)\C=C\C1=CC=C(Br)C=C1F
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 4-Bromo-2-fluorocinnamic acid, 98%, Thermo Scientific™
CAS: 149947-19-3 Molecular Formula: C9H6BrFO2 Molecular Weight (g/mol): 245.047 MDL Number: MFCD00143267 InChI Key: SVJGKQYKYNDYMU-DUXPYHPUSA-N Synonym: 4-bromo-2-fluorocinnamic acid,2e-3-4-bromo-2-fluorophenyl prop-2-enoic acid,e-3-4-bromo-2-fluorophenyl acrylic acid,trans-4-bromo-2-fluorocinnamic acid,3-4-bromo-2-fluoro-phenyl-acrylic acid,rarechem bk hw 0191,4-bromo-2-fluorocinnamicacid,pubchem3229,e-3-4-bromo-2-fluorophenyl prop-2-enoic acid PubChem CID: 688314 IUPAC Name: (E)-3-(4-bromo-2-fluorophenyl)prop-2-enoic acid SMILES: C1=CC(=C(C=C1Br)F)C=CC(=O)O