man

Test

CAS RN 15074-54-1

ClOPClClO

CAS RN 15074-54-1

IUPAC Name: 2-chlorophenyl chlorophosphonochloridate
Synonyms: 2-chlorophenyl chlorophosphonochloridate
Molecular Weight (g/mol): 245.42
Molecular Formula: C6H4Cl3O2P
InChi Key: VLDPXPPHXDGHEW-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1OP(Cl)(Cl)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results