man

Test

CAS RN 151945-84-5

H3COClF

CAS RN 151945-84-5

IUPAC Name: 1-(4-chloro-3-fluorophenyl)ethan-1-one
Synonyms: 1-(4-chloro-3-fluorophenyl)ethanone
Molecular Weight (g/mol): 172.58
Molecular Formula: C8H6ClFO
InChi Key: ATZHNDSADJDUPJ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(Cl)C(F)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results