CAS RN 15199-43-6
CAS RN 15199-43-6
IUPAC Name:
di(²H₃)methyl sulfate
Synonyms:
di(²H₃)methyl sulfate
Molecular Weight (g/mol):
132.16
Molecular Formula:
C2H6O4S
InChi Key:
VAYGXNSJCAHWJZ-WFGJKAKNSA-N
SMILES:
[2H]C([2H])([2H])OS(=O)(=O)OC([2H])([2H])[2H]