man

Test

CAS RN 15260-10-3

H3COHHNOOOOCH3CH3CH3

CAS RN 15260-10-3

IUPAC Name: (2S,3R)-3-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}butanoate
Synonyms: (2S,3R)-3-(benzyloxy)-2-[(tert-butoxycarbonyl)amino]butanoate
Molecular Weight (g/mol): 308.36
Molecular Formula: C16H22NO5
InChi Key: CTXPLTPDOISPTE-YPMHNXCESA-M
SMILES: C[C@@H](OCC1=CC=CC=C1)[C@H](NC(=O)OC(C)(C)C)C([O-])=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results