man

Test

CAS RN 15295-77-9

OONH2HBr

CAS RN 15295-77-9

IUPAC Name: 3-aminooxolan-2-one hydrobromide
Synonyms: HSLs hydrobromide
Molecular Weight (g/mol): 182.02
Molecular Formula: C4H8BrNO2
InChi Key: MKLNTBLOABOJFZ-UHFFFAOYNA-N
SMILES: Br.NC1CCOC1=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)