man

Test

CAS RN 1530-03-6

CH3

CAS RN 1530-03-6

IUPAC Name: (1-phenylpropyl)benzene
Synonyms: 1,1-diphenylpropane
Molecular Weight (g/mol): 196.29
Molecular Formula: C15H16
InChi Key: BUZMJVBOGDBMGI-UHFFFAOYSA-N
SMILES: CCC(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results