accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 154230-29-2

HOBOHCl(E)

CAS RN 154230-29-2

IUPAC Name: [(1E)-2-(4-chlorophenyl)ethenyl]boronic acid
Synonyms: (1E)-2-(4-chlorophenyl)ethenylboronic acid
Molecular Weight (g/mol): 182.41
Molecular Formula: C8H8BClO2
InChi Key: HWSDRAPTZRYXHN-AATRIKPKSA-N
SMILES: OB(O)\C=C\C1=CC=C(Cl)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products