man

Test

CAS RN 1547-29-1

HNOF

CAS RN 1547-29-1

IUPAC Name: 3-fluoro-1,2-dihydropyridin-2-one
Synonyms: 3-fluoro-1H-pyridin-2-one
Molecular Weight (g/mol): 113.09
Molecular Formula: C5H4FNO
InChi Key: CISUDJRBCKKAGX-UHFFFAOYSA-N
SMILES: FC1=CC=CNC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results