man

Test

CAS RN 156772-60-0

NBrBrF

CAS RN 156772-60-0

IUPAC Name: 2,5-dibromo-3-fluoropyridine
Synonyms: 2,5-dibromo-3-fluoropyridine
Molecular Weight (g/mol): 254.88
Molecular Formula: C5H2Br2FN
InChi Key: QEENLQKOTDHQEW-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CN=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results