man

Test

CAS RN 156886-85-0

H3CHNHNHNHNCH3HClHClHClHCl

CAS RN 156886-85-0

IUPAC Name: 3,7,11,15-tetraazaheptadecane tetrahydrochloride
Synonyms: 3,7,11,15-tetraazaheptadecane tetrahydrochloride
Molecular Weight (g/mol): 390.26
Molecular Formula: C13H36Cl4N4
InChi Key: LAWBSOKBIROCQP-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.Cl.CCNCCCNCCCNCCCNCC
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results