accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 15743-20-1

AlOOH2COOH2COOH2C

CAS RN 15743-20-1

IUPAC Name: aluminium(3+) tris(prop-2-enoate)
Synonyms: aluminium(3+) triacrylate
Molecular Weight (g/mol): 240.15
Molecular Formula: C9H9AlO6
InChi Key: LIQDVINWFSWENU-UHFFFAOYSA-K
SMILES: [Al+3].[O-]C(=O)C=C.[O-]C(=O)C=C.[O-]C(=O)C=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products