CAS RN 1633044-56-0
CAS RN 1633044-56-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express FIIN-2, MedChemExpress
MedChemExpress FIIN-2 is an irreversible inhibitor of FGFR with an IC50 of 3.1, 4.3, 27, and 45 nM for FGFR1, FGFR2, FGFR3 and FGFR4, respectively.
